Screw caps and septa for Reacti-Vial™ and screw cap septum vials
Supplier: Thermo Fisher Scientific
HYPETS-13218EA 64.7 EUR
HYPETS-13218 HYPETS-13215 HYPETS-13216 HYPETS-13219
Screw caps and septa for Reacti-Vial™ and screw cap septum vials
Closures Chromatography Vial Closures
These open top screw caps provide an inert, air-tight seal and direct puncture access to samples with a syringe needle.
  • Fits into Reacti-Vial™ small reaction vials
  • Used with PTFE/rubber laminated discs