Leupeptin hemisulphate
Supplier: VWR Chemicals
J580-25MGEA 183 EUR
J580-25MG J580-5MG
Leupeptin hemisulphate
Proteins and Peptides
A reversible inhibitor of cysteine proteases and serine proteases resembling trypsin. Inhibits trypsin, plasmin, papain, kallikrein, thrombin, and cathepsin A and B.