E-64

Supplier: Thermo Fisher Scientific
J62933.MBEA 534 EUR
J62933.MB J62933.X0
E-64
Proteins and Peptides
  • Light Sensitive

CAS No.: 66701-25-5
Molecular Formula: C15H27N5O5
Formula Weight: 357.40
Storage Temperature: -30°C to -10°C
Physical Form: Powder
Appearance: White to off-white
Solubility: Soluble in DMSO (25mg/ml), or water (20mg/ml)
Beilstein Reference: 1405664
Optical Rotation: +24° (c=1 in 1N HCl)
MDL No.: MFCD00080261

Selective and irreversible cysteine protease inhibitor
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR