Beakers, low form
Supplier: Kartell
plastilab®
KART1538EA 395 EUR
KART1538 KART1537 KART1539 KART1534 KART1536 KART1535
Beakers, low form
Beakers
PFA, translucent, with spout.
  • Space-saving stackable design
  • High thermal resistance from −250 to +270 °C
  • Autoclavable, easy to clean
Embossed graduations are easy to read and resistant to wear from repeated use.