Magnetic stirring bars, PTFE, oval
Supplier: Kartell
KART683EA 17.3 EUR
KART683 KART684 KART685 KART686 KART687 KART688 KART689
Magnetic stirring bars, PTFE, oval
Stirrers Magnetic Stirring Bars
These oval shaped PTFE stirring bars are ideal for containers or bottles with round bottom.
  • Suitable for food stuff