Magnetic stirring bars, triangular
Supplier: Kartell
KART781EA 10.6 EUR
KART781 KART782 KART783 KART780
Magnetic stirring bars, triangular
Stirrers Magnetic Stirring Bars
PTFE coated, triangular magnetic stirring bars.
  • Scraping action ideal for dissolving solids and mixing sediments
  • Angled surfaces create high turbulence even at low speeds
  • Suitable for food stuff