Centrifuge tubes, conical bottom
Supplier: Kartell
plastilab®
KART84011EA 364 EUR
KART84011 KART84012 KART84013 KART84014 KART84010 KART84008 KART84009 KART84015 KART84006 KART84007
Centrifuge tubes, conical bottom
Tubes Centrifuge Tubes
PP, translucent.
  • Very good chemical resistance
  • Very good temperature resistance
  • Graduated
Conical bottom with tight screw cap.