Centrifuge tubes, PP, with screw cap
Supplier: Kartell
Dispolab®
KART8830304EA 354 EUR
KART8830304 KART8830310 KART88304-03 KART88303/03 KART88303/06
Centrifuge tubes, PP, with screw cap
Tubes Centrifuge Tubes
PP with quick closure PE screw cap.
  • Leak-proof
  • Can be centrifuged up to 4000×g
  • Cap is designed to avoid any contamination due to drops of liquid on the inner surface