Microplate fluorometers, Fluoroskan™
Supplier: Thermo Scientific
LABL5200111EA 20570 EUR
LABL5200111 LABL5200110 LABL5200112
Microplate fluorometers, Fluoroskan™
Microplate Readers
The Thermo Scientific™ Fluoroskan™ is a compact and robust microplate fluorometer offering simplicity and reliability for increased efficiency of workflows. Featuring excellent optical performance, the Fluoroskan Microplate Fluorometer is ideal for life science research applications such as fluorometric protein and enzyme studies, molecular interaction studies, nucleic acid quantification, as well as reporter gene, fluorometric kinase, immunological, and cell-based assays.
  • High sensitivity for both top and bottom reading
  • Onboard dispensers for precise initiation of kinetic reactions
  • Powerful and easy to use Thermo Scientific™ SkanIt™ software
The use of fibreless direct illumination optics allows for both top and bottom reading enables high sensitivity, a wide dynamic range, and accurate and precise results for 6- to 384-well plates.
The Fluoroskan™ Microplate Fluorometer can be equipped with up to three reagent dispensers* for use during fast kinetic assays, such as Ca₂+ flux assays. The onboard dispensers support simultaneous dispensing and reading, enabling the monitoring of fast kinetic measurements from the very start of the reaction. Very low dead volume and backflush of reagents support conservation of reagents.
With a visual workflow setup, SkanIt™ software is quick and easy to learn. It offers easy data analysis (with a comprehensive selection of built-in calculations, including blank subtraction, standard curve creation, classification, quality control, kinetic analysis) and exporting capabilities. Each user can install SkanIt™ software on his/her own computer to set up assays and analyze data.
Ordering information: Includes SkanIt™ PC Software and the following filter pairs (excitation/emission): 355 nm/460 nm, 485 nm/538 nm. An extensive selection of additional filters is available upon request.
Caution: * Upgrade to third dispenser must be performed by service technician