Screw caps for screw neck vials, N 8
Supplier: MACHEREY-NAGEL
MANA702066EA 27.9 EUR
MANA702066 MANA702067 218-1270 MANA70245 MANA702437 MANA70250 MANA702068 MANA702069
Screw caps for screw neck vials, N 8
Closures Chromatography Vial Closures
The suitable screw closures are ready assembled.
  • Septa made from economical Red Rubber/FEP, high quality silicone/PTFE or from virgin PTFE
  • Silicone/PTFE septa are also slitted