Crimp caps for crimp neck vials, N 20
Supplier: Macherey-Nagel
MANA70233EA 56 EUR
MANA70233 218-1271 MANA70234 MANA70235 MANA70234.8 MANA702839 MANA70234.9 MANA702837 MANA702838 MANA70236.1 MANA702835 MANA702833 MANA702834 MANA702799 218-1272 MANA702775 MANA702112 MANA702773 MANA702928.9 MANA70266 MANA70267 MANA702093 MANA702071 MANA702094 MANA702808 MANA702929 MANA702927 MANA702829 MANA702804 MANA702805 MANA702129 MANA702128 MANA70240 MANA702145 MANA70234.10
Crimp caps for crimp neck vials, N 20
Closures Chromatography Vial Closures
Broad range of different crimp caps with mostly assembled septa, stoppers and PE caps are available for the crimp neck vials.
  • Center hole caps
  • Aluminium pressure release caps with special perforation for burst protection
  • Center tear off caps and complete tear off caps
  • Bi-metal crimp caps and magnetic crimp caps for autosamplers with magnetic transportation system
  • Silicone / PTFE septa (required above 120 °C) and Butyl / PTFE septa in different versions