Crimp caps for crimp neck vials, N 11
Supplier: Macherey-Nagel
MANA702001EA 13.5 EUR
MANA702001 MANA70288 MANA702175 MANA70256 MANA70239 MANA702995.3 MANA702879 MANA70231.3 MANA702730.1 MANA70288.3 MANA70288.2 MANA70231.2 MANA702823 MANA702801 MANA702730.3 MANA70288.1 MANA702995.1 MANA702995.2 MANA702730.2 MANA702730 MANA702401 MANA702995 MANA70284 MANA70231.1
Crimp caps for crimp neck vials, N 11
Closures Chromatography Vial Closures
Ready-to-use crimp caps with assembled septa and plain crimp caps are available for the crimp neck vials.
  • Broad range of aluminium crimp and magnetic crimp closures with different septum materials and cap colours
  • Magnetic version for CTC PAL autosampler or identical constructions
  • Large choice of septa materials