Test papers, qualitative, Bismuth
Supplier: Macherey-Nagel
MANA90733EA 43.6 EUR
MANA90733
Test papers, qualitative, Bismuth
Environmental Test Kits
This test paper allows the rapid detection of Bismuth. In the presence of Bismuth ions (Bi³⁺) it shows an orange-red discolouration on a pale-yellow background.
  • Colour change: Pale-yellow to orange-red
Limit of sensitivity: 60 mg/L Bi³⁺ (Nitric acid >3% leads to reduced sensitivity)
Delivery information: Pack of 200 strips 20×70 mm.