Colorimetric rapid tests, nitrite, VISOCOLOR®
Supplier: Macherey-Nagel
VISICOLOR®
MANA931044EA 52.4 EUR
MANA931044 MANA931244 MANA920163 MANA920063 MANA935066
Colorimetric rapid tests, nitrite, VISOCOLOR®
Environmental Test Kits
The VISOCOLOR® testkits are devided into VISOCOLOR® alpha, ECO and HE. Every product range offering its specific strenght to provide a tailored test for each and every user and application.
  • Portable
  • Environmentally friendly
  • Convenient handling
  • Application-oriented measuring ranges
In surface waters nitrite ions are generally present in low concentrations. Their presence in ground water is less common. In waste waters nitrite frequently occurs, even in fairly high concentrations.