Colorimetric rapid tests, chlorine, VISOCOLOR®
Supplier: Macherey-Nagel
VISICOLOR®
MANA936220.1EA 186 EUR
MANA936220.1 MANA931090 MANA920115 MANA920015 MANA931290 MANA931035 MANA931016 MANA931015 MANA931235 MANA931216 MANA935019 MANA931215 MANA931217 MANA931219
Colorimetric rapid tests, chlorine, VISOCOLOR®
Environmental Test Kits
The VISOCOLOR® testkits are devided into VISOCOLOR® alpha, ECO and HE. Every product range offering its specific strenght to provide a tailored test for each and every user and application.
  • Portable
  • Environmentally friendly
  • Convenient handling
  • Application-oriented measuring ranges
The addition of chlorine to swimming pools, water reservoirs and water mains is an approved procedure to rid the water of germs. With the correct dose harmful microorganisms are destroyed, many impurities removed and the growth of algae is prevented. However it is imperative that the chlorine content is regularly checked, since excessive chlorine not only impairs the smell and taste of the water but can be hazardous.
One distinguishes between free chlorine and bound chlorine (chloroamines); the sum of both is called total chlorine.