Photometric tube tests, NANOCOLOR® ammonium
Supplier: Macherey-Nagel
NANOCOLOR®
MANA985008EA 85.7 EUR
MANA985008 MANA985002 MANA985005 MANA985006 MANA985003 MANA985004
Photometric tube tests, NANOCOLOR® ammonium
Environmental Test Kits
A range of analytical reagents for photometric determination of ammonium in water analysis using a pre-dosed tube.
  • Measurement directly in the test tube, saving time and reducing exposure to chemical reagents
  • Bar code cuvette identification
  • Precisely pre-dosed reagents in 16 mm (Ext.Ø) tubes
Tests that are particularly suitable for the NANOCOLOR® photometers but can be used with other photometers.