Collection containers, Corning®
Supplier: Corning
Corning®
MDTC91-200-39EA 74.71 EUR
MDTC91-200-39 MDTC91-100-30 MDTC91-001-MB MDTC91-200-42 MDTC91-200-20 MDTC91-200-10 MDTC91-200-43 MDTC91-200-01 MDTC91-200-45 MDTC91-200-02 MDTC91-200-47 MDTC91-200-36 MDTC91-200-48 MDTC91-200-05 MDTC91-200-82 MDTC91-200-83 MDTC91-200-41
Collection containers, Corning®
Bags Media Bags
Corning® collection containers are fabricated to match specific application requirements, and are available in a variety of sizes and connector configurations.
  • Single-use bags
  • Material ULDPE
  • Designed to be practical and cost-effective alternatives to rigid-walled containers
  • Ideal for biopharmaceutical applications
Unless otherwise noted in container port descriptions, all port tubing components are chemically resistant, heat sealable flexible tubing.
Port 1: 60 L × ¹/₂ Int.Ø ×³/₄ Ext.Ø tubing, Male MPX connector, Female MPC cap, pinch clamp.
Port 2: 40 L × ¹/₈ Int.Ø ×¹/₄ Ext.Ø tubing, Female Luer with ⁵/₃₂" Barb, Swabable valve, Non-breather Female Luer cap.
Port 3: 60 L × ¹/₂ Int.Ø ×³/₄ Ext.Ø tubing, 1,5" Tri-clover with ¹/₂" Barb, pinch clamp