Sterilisers, SFplus
Supplier: Memmert
MEMMSF450PLUSEA 5750 EUR
MEMMSF450PLUS MEMMSF75PLUS MEMMSF55PLUS MEMMSF30PLUS MEMMSF160PLUS MEMMSF110PLUS MEMMSF750PLUS MEMMSF260PLUS
Sterilisers, SFplus
Sterilisers Hot Air Sterilisers
These sterilisers comply with all relevant standards and regulations for medical products, and guarantee reliable program sequences, irrespective of the load and chamber volume. With the SetpointWAIT function validation of this hot air steriliser is possible anytime. The SetpointWAIT function ensures that the program only continues when the set point temperature has been reached. Precisely maintaining the sterilisation time and thus completely killing off even highly resistant microorganisms is, therefore, guaranteed at all times, irrespective of the sterilisation load and chamber volume. This feature can also be used with additional, freely positionable PT100 temperature sensors. In this case, sterilisation only begins when the set point temperature has been reached at all measurement points – that is, in the load. For special applications such as depyrogenisation, this hot air steriliser is absolutely ideal. SFplus models are equipped with TwinDISPLAY.
  • Forced air circulation
  • Fully insulated stainless steel door with 2-point locking
  • Auto diagnostic system for fault analysis
  • Program stored in case of power failure
  • Internal data logger with a storage capacity of at least 10 years
  • AtmoCONTROL software for reading out, managing and organising the data logger via ethernet interface (temporary trial version can be downloaded)
  • Digital backwards counter with target time setting, adjustable from 1 minute to 99 days
  • ControlCOCKPIT for operation and adjustment of all parameters
  • Double over-temperature protection
Material: Corrosion-resistant, easy to clean interior made of stainless steel, reinforced by deep drawn ribbing with integrated and protected large area heating on four sides. Rear zinc plated steel housing.
TwinDISPLAY includes HeatBALANCE function, AutoSAFETY function, user-ID, USB port, individual limits for over-/under-temperature, over-/under-humidity. Ethernet interface on the rear of the appliance for reading the protocol log and for uploading and implementing programs and for online logging.
Digital backwards counter with target time setting, adjustable from 1 minute to 99 days. AtmoCONTROL software on a USB stick for programming, managing and transferring programs via ethernet interface or USB port. German, English, French, Spanish language settings available on the ControlCOCKPIT.
Certifications: Electronic temperature monitoring TWW/TWB (protection class 3.1 or 2 resp. 3.3 for units with active cooling) and mechanical temperature limiter TB (protection class 1) according to DIN 12 880. Medical device class IIa according to European Medical Devices Directive 93/42/EEC.
Ordering information: Supplied without shelves, please order required shelves, stainless steel grids or trays separately. For all chamber volumes > 53 litres, a minimum of 2 shelves or grids must be ordered in addition.