Hose Clamps, Adjustable, Stainless Steel
Supplier: Avantor Fluid Handling
Masterflex®
MFLX06403-14EA 38.6 EUR
MFLX06403-14 MFLX06403-25 MFLX06403-15 MFLX06403-16 MFLX06403-21 MFLX06403-22 MFLX06403-11 MFLX06403-12 MFLX06403-23 MFLX06403-24 MFLX06403-13
Hose Clamps, Adjustable, Stainless Steel
Clamps Tubing and Sanitary Clamps
Ideal for securely clamping tubes in place.
  • Resists corrosion
The band-type worm drive clamp features stainless steel strap and screw housing with a stainless steel screw.