Masterflex® Ismatec® Replacement Cartridges and Adapters, Avantor®
Supplier: Avantor Fluid Handling
Masterflex®
MFLX78016-98EA 70.3 EUR
MFLX78016-98 MFLX07624-64 MFLX78016-96 MFLX07624-96 MFLX78001-97 MFLX07618-67 MFLX78001-95 MFLX07618-69 MFLX78001-96 MFLX07618-73 MFLX78001-90 MFLX07618-75 MFLX78001-91 MFLX07618-65 MFLX07618-71
Masterflex® Ismatec® Replacement Cartridges and Adapters, Avantor®
Pumps Pump Heads Peristaltic Pump Heads
Choose new cassettes based upon material of construction, tubing type and pressure capabilities.
  • Material of construction standard POM-C or optional PVDF
There are two different designs of cartridge. The Click-N-Go design allows for tubing to be used in systems that have up to 1 bar of back pressure exerted on the tubing. The Pressure Lever design allows for system pressures up to 1.5 bar.