Masterflex® Hose Clamps, Nylon, Avantor®
Supplier: Avantor Fluid Handling
Masterflex®
MFLX98766-42EA 107 EUR
MFLX98766-42 MFLX98766-31 MFLX98766-32 MFLX98766-33 MFLX98766-34 MFLX98766-35 MFLX98766-36 MFLX98766-37 MFLX98766-38 MFLX98766-39 MFLX98766-41
Masterflex® Hose Clamps, Nylon, Avantor®
Clamps Tubing and Sanitary Clamps
Secure tubing to connectors, pumps, glassware, and filtration devices.
  • Use to fasten pressure, vacuum, and liquid tubing lines
  • Nylon is corrosion-proof, chemical resistant, and non-conductive
  • Works with all types of plastic and rubber tubing
Reusable clamp squeezes shut for a leakproof seal—press the sides to release the clamp. Order the size that suits your needs, or the handy kit to handle multiple projects.