Microplate handling system, StakMax®
Supplier: Molecular Devices
MLDV0200-6017EA 2870 EUR
MLDV0200-6017 MLDV0310-5102 MLDV5005331 MLDV0200-6187 MLDV0310-5101 MLDVSTAKMAX MLDV0200-6016
Microplate handling system, StakMax®
Microplate Readers
The StakMax® microplate handling system is a compact benchtop microplate stacker that works with microplate readers and microplate washers to give walk-away automation for up to 50 microplates.
  • Integrates with microplate readers and AquaMax® washers for improved process flow and higher throughput
  • Flexible operation using powerful SoftMax® Pro software
  • Barcode reader can be added to enable barcode identification of plates
  • Minimises plate movement time, while handling the microplates gently, eliminating liquid spillage