DNA dyes, EZ-Vision®
Supplier: VWR Chemicals
EZ-Vision®
N313-KITEA 228 EUR
N313-KIT N650-KIT N472-KIT
DNA dyes, EZ-Vision®
Stains Electrophoresis Stains
EZ-Vision DNA dye as loading buffer provides a non mutagenic, environmentally friendly alternative to ethidium bromide for instantaneous fluorescent DNA band visualisation.
  • Easy and convenient for instant results, low background
  • Completely eliminates the need for ethidium bromide
  • No post electrophoresis staining or destaining, very broad emission spectra with a peak near 450 nm that works with most existing filters for gel documentation
Each version contains the same non hazardous fluorescent DNA dye and loading buffer that can be run on a gel and visualised immediately on a UV transilluminator. EZ-Vision One is one tracking dye migrating at approximately 10 bp in a 1% agarose gel. EZ-Vision Two is two tracking dyes migrating at approximately 4000 bp and 400 bp in a 1% agarose gel. EZ-Vision Three is three tracking dyes migrating at approximately 4000 bp, 400 bp and 10 bp in a 1% agarose gel. This dye requires no special handling or disposal and will not incur hazardous shipping, storage or waste charges.
Delivery information: Contains 1, 2 or 3 coloured tracking dyes in 6X loading buffer.