Double spatulas, Chattaway type, nickel
Supplier: Nickel-Electro
231-0207EA 12.8 EUR
231-0207 NICK2216 NICK2202 231-0164 231-0162 231-0163 231-0160 231-0209
Double spatulas, Chattaway type, nickel
Spatulas
Multipurpose spatula made of pure nickel (99,6%) for a wide range of uses. One end is flat and spatula-shaped, the other end is rounded and turned up at 45°. Available in light or medium weight design.
  • Excellent chemical resistance
  • Excellent thermal resistance
  • Quick and easy to clean