Retort bases, with M10 thread
Supplier: Nickel-Electro
NICK6891EA 24.6 EUR
NICK6891 NICK6890 NICK6893 NICK6892
Retort bases, with M10 thread
Retort Stands
Pressed steel finished in matt black textured acrylic paint, threaded M10 × 1,5 mm.
Ordering information: Retort rods are not included, they can be ordered separately under "Complementary products".