Spoon spatulas, micro
Supplier: Nickel-Electro
NICK8065EA 247 EUR
NICK8065 NICK8064
Spoon spatulas, micro
Spatulas
Pure nickel (99,6%).
  • One end flattened, the other end spoon-formed
  • Good chemical resistance
  • Very good thermal resistance
  • Quick and easy to clean