Cleanroom hoods, BioClean-D™, Clean-Tough™, longer length
Supplier: Ansell-Nitritex
BioClean-D™ Clean Tough™
NITRBDHD-LEA 250 EUR
NITRBDHD-L NITRS-BDHD-L
Cleanroom hoods, BioClean-D™, Clean-Tough™, longer length
Disposable Garments
These disposable hoods are made from antistatic lightweight and comfortable Clean-Tough™ material. They features a three-piece design to ensure a perfect fit. The extra-long yoke ensures maximum coverage and avoids contamination when entering the controlled environment when worn in combination with a coverall with collar.
  • Elasticated face opening with reinforced edges
  • Lightweight, low linting and hard-wearing material
  • Available as sterile or non sterile version
  • A unique barcode on the outer carton (ITF-14) allows automatic data entry into stock control systems
Suitable for use in Class 10, ISO 4, GMP Grade A environments.
Certifications: EN ISO 13034 (Type PB[6]), EN 14325, EN 1149-5, EN 530, EN 7854, EN 6530, EN 13934-1, EN 9073-4, EN 863, EN 13935-2; Cat. III
Manufactured in a facility holding ISO 9001:2008 and 89/686/EEC Article 11b certifications.
Sterile model: Sterilised in accordance with ISO 11137-1:2006.
Packaging: Sterile version: one hood per inner bag, 20 inner bags per sealed outer bag; six outer bags per lined carton (120 pieces).
Non sterile version: 20 pieces per sealed double plastic bag; one inner bag per sealed outer bag; six outer bags per lined carton (120 pieces).