Cleanroom overalls, Bioclean-R™
Supplier: Ansell-Nitritex
NITRSTC002-A40EA 0 EUR
NITRSTC002-A40 NITRSTC002-A30 NITRSTC002-A20 NITRSTC002-A10 NITRSTC002 NITRSTC001-A20 NITRSTC001-A10 NITRSTC001-A40 NITRSTC001-A30 NITRSTC003-A20 NITRSTC003-A10 NITRSTC003-A40 NITRSTC003-A30
Cleanroom overalls, Bioclean-R™
These overalls with collar are made of MT811 fabric consisting of 99% polyester and 1% carbon.
STC001
- Raglan sleeves
- Centre fastening zip with YKK slider
- Elastic back
- Stud wrist and ankles
STC002
- Suitable for gamma irradiation
- Raglan sleeves
- Centre fastening zip with YKK slider
- No elastic back
- Stud neck, wrists and ankles
STC003
- Raglan sleeves
- Centre fastening zip with YKK slider
- Elastic back
- Lycra wrists
- Stud ankles
Suitable for use in ISO class 4, 5, 6 and 7 cleanrooms.
Weight: 113 g/m²