Guardian 5000 magnetic hotplate stirrers
Supplier: Ohaus
Guardian 5000
OHAU30500563EA 595 EUR
OHAU30500563 OHAU30500553 OHAU30500523
Guardian 5000 magnetic hotplate stirrers
Stirrers Magnetic Hotplate Stirrers
Reliable magnetic hotplate stirrer with exceptional safety features for everyday use.
  • SafetyHeat™ system protects the laboratory
  • Powerful, consistent stirring performance
  • Easy to read LCD display
  • Designed for durability and reliability with SmartHousing™
Guardian 5000™ magnetic hotplates stirrers are designed for safety and simplicity in all research, academic and industrial applications.
The design ensures proper mixing and temperature control of the top plate or your sample when using the optional temperature probe.
Heating models include a Single Point Calibration feature for multiple setpoints. Choice of models with differing plate sizes to suit different everyday heating and stirring applications.