Centrifugal devices for RNA and DNA extraction, NAB Nanosep®
Supplier: Pall Laboratory
PALLODNABC33EA 36.8 EUR
PALLODNABC33 PALLODNABC34
Centrifugal devices for RNA and DNA extraction, NAB Nanosep®
Filters Centrifugal Filters
The Pall Nanosep device for nucleic acid binding (NAB) incorporates a dual layer silica-based quartz glass fiber media to allow for efficient binding of DNA and RNA, while providing smooth flow and rapid processing of samples.
  • Rapid processing of samples
  • Silica-based quartz glass fiber media that allows efficient binding of DNA and RNA
  • Constructed of low-binding polypropylene
  • Ultrasonically welded seals prevent by passor seal failure
  • Fits standard centrifuge rotors that accept 1,5 ml tubes
This media offers researchers the flexibility to purify plasmid DNA, genomic DNA or total RNA from a variety of starting materials: a single column for multiple applications. The new NABNanosep device offers flexibility without sacrificing on quantity or quality of the nucleic acid. The Pall NAB Nanosep device is a multipurpose centrifugal device providing flexibility in applications, supported by comprehensive protocols for sample processing.
The dual filter media is made from binder-free, silica-based glass fiber and comes in a polypropylene housing. Has a binding capacity of up to 160 μg pure RNA, 69 μg pure genomic DNA, 11,5 μg pure plasmid DNA.
Has a minimum elution volume of 40 μl and capable of base pair binding from 50 bp.