WFI samplers, Avantor®
Supplier: Avantor Fluid Handling
PAWBTS41600AT018EA 396 EUR
PAWBTS41600AT018 PAWBTS41200AT036 PAWBTS41600AT036 PAWBTS41200AT018
WFI samplers, Avantor®
Samplers Fluid Samplers
WFI Samplers are an easy and convenient solution for obtaining perfect fluid samples from a WFI water drop.
  • Made from AdvantaFlex tubing
  • PP fitting is inert and strong
  • Silicone tubing and gasket: unlike TPE materials, silicone will not contribute to TOC counts
  • Gamma-irradiation eliminates additional bio-burden concerns
The fittings utilise the SterilEnz-II/AT fitting platform, which is pre-gasketed. This means a quick fit to the water system or equipment without further contamination or gasket hassles.
Tubing size (Int.Ø×Ext.Ø): 6,35×11,11 mm
Certifications: USP 88 Class VI Biological Test for Plastics. USP 87 MEM Elution Test. USP 661.
Certified free from any animal derived components or processes.
Packaging: Each WFI Sampler is double poly-pouched for cleanroom use and irradiated.