Gas washing bottles, with filter candle, VitraPOR®
Supplier: ROBU
VitraPOR®
ROBU42133EA 68.45 EUR
ROBU42133 ROBU42132 ROBU42131 ROBU42130 ROBU42134 ROBU42092 ROBU42091 ROBU42090 ROBU42094 ROBU42093
Gas washing bottles, with filter candle, VitraPOR®
Bottles Gas Washing Bottles
They are made of borosilicate glass 3.3 and the versatile and recyclable material offers good properties for the most diverse fields.
  • Resistant to chemicals such as solvents, acids and alkalis
  • Very good temperature resistance
  • With screw cap and silicone washer