X-ray and metal detectable jugs, PP, graduated
Supplier: Sampling Systems
SAMPM1044B-1000EA 86.2 EUR
SAMPM1044B-1000 SAMPM1044B-500
X-ray and metal detectable jugs, PP, graduated
Beakers
X-ray and metal detectable jugs are perfect for both laboratory and production work. They are available in 2 sizes. The bright blue colour makes the jugs easy to spot if they get left in the product.
  • Robust PP construction
  • Excellent chemical resistance
  • Non sterile
  • Full batch traceability
To ensure the cleanliness of the jugs, they are manufactured and packed in a cleanroom. The robust nature of these jugs means that they can give many years of service or for more demanding applications. They can be used as a single use disposable product straight from the bag.
Ideal for use in food, pharma, chemical industries and use with powders, granules and pastes.
Certifications: These PP jugs conform to all the latest standards including FDA 177.1520, EU 10/2011, EC 1935/2004 and are BSE/TSE free. Batch certificates are available.
Packaging: Supplied individually bagged.