HPLC bottles with conical base GL 45, DURAN®
Supplier: DWK Life Sciences
DURAN®
215-4547EA 1484.06 EUR
215-4547 215-4546 215-4545 215-4544 215-4543
HPLC bottles with conical base GL 45, DURAN®
Bottles HPLC Bottles
DURAN®, borosilicate glass 3.3, clear, with blue PP screw cap.
  • Central cavity can accommodate magnetic stir bars for improved mixing
  • Side vent or drain hole in the base for ultrasonic bath degassing
  • Durable white enamel volume graduations
  • Autoclavable at 121 °C or 134 °C
Applications include mobile phase reservoirs for HPLC, reagent supply to liquid handling systems. DURAN® reservoirs have been specially designed for the storage and delivery of high performance liquid chromatography mobile phases and solvents. Reservoirs have one cavity for the filter to allow the most efficient delivery of mobile phase to HPLC analysers.
A safer and more convenient alternative to flat bottomed glass bottles, the conical bottom reservoirs allow a greater recovery of solvent, without having to tilt reservoir and risk a solvent spill. The larger reservoirs allow longer, uninterrupted analyses. These are especially useful during a weekend or lengthy chromatographic analysis when there might be a risk of running out of solvents