Dropping funnels, cylindrical, without pressure balance, DURAN®
Supplier: DWK Life Sciences
SCOT241244403EA 160.38 EUR
SCOT241244403 SCOT241242402 SCOT241242008 SCOT241245605 SCOT241243604 SCOT241242505 SCOT241244609 SCOT241243707
Dropping funnels, cylindrical, without pressure balance, DURAN®
Funnels Dropping Funnels
DURAN®, borosilicate glass 3.3, clear, with glass key.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Cylindrical shape. With standard ground joint cone and socket. Clearly visible divisions for approximate volume reading.
DIN ISO 4800.
* non-DIN ISO size