Dropping funnels, cylindrical, with pressure balance, DURAN®
Supplier: DWK Life Sciences
SCOT241252506EA 151.68 EUR
SCOT241252506 SCOT241252009 SCOT241253605 SCOT241252403 SCOT241254404 SCOT241254601 SCOT241253708
Dropping funnels, cylindrical, with pressure balance, DURAN®
Funnels Dropping Funnels
DURAN®, borosilicate glass 3.3, clear, with glass key.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Standard ground joint cone and socket, with divisions for approximate volume reading.
* non-DIN ISO size
Certifications: DIN ISO 4800