Aspirator bottles, with tube connector and drain cock, DURAN®
Supplier: DWK Life Sciences
DURAN®
SCOT247025401EA 156.74 EUR
SCOT247025401 SCOT247026303 SCOT247024405 215-1001
Aspirator bottles, with tube connector and drain cock, DURAN®
Bottles Aspirators
DURAN®, borosilicate glass 3.3, clear.
  • Very good chemical resistance
  • High thermal resistance, autoclavable
  • Minimal thermal expansion giving relatively high resistance to temperature changes
With ground tube connector NS 29/32 and ground drain cock. Neck with ground sleeve and ground glass stopper.
Certifications: DIN 12037