Aspirator bottles, narrow mouth, with stopcock, DURAN®
Supplier: DWK Life Sciences
SCOT247037309EA 298.02 EUR
SCOT247037309 215-1008 SCOT247035402 SCOT247036304
Aspirator bottles, narrow mouth, with stopcock, DURAN®
Bottles Aspirators
DURAN®, borosilicate glass 3.3, clear.
  • Very good chemical resistance
  • High thermal resistance, autoclavable
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Neck with DIN thread GL 45, tubulator with GL 32. Complete with screw connection cap, silicone seal and also stopcock with PTFE spindle. Dosing of liquids is possible via stopcock.
With printed retrace code (batch identification).