Woulff bottles, DURAN®
Supplier: DWK Life Sciences
189-1110EA 198.24 EUR
189-1110 SCOT247104401 SCOT247107304
Woulff bottles, DURAN®
Bottles Woulff Bottles
DURAN®, borosilicate glass 3.3, clear.
  • With bottom tubulature NS 19/26
  • Very good temperature resistance
  • Very good chemical resistance
Bottle with 3 grounded necks.
Certifications: DIN 12481.