Stopcock, single way, taper 1:5, DURAN®
Supplier: DWK Life Sciences
SCOT286457002EA 227.2 EUR
SCOT286457002 SCOT286456409 SCOT286456006 SCOT286457208 SCOT286455404 SCOT286455601
Stopcock, single way, taper 1:5, DURAN®
Stopcocks
DURAN®, borosilicate glass 3.3, clear, polished, without key.
  • Very good chemical resistance
  • Very good temperature resistance
  • For PTFE keys
Straight bore.