CHO cell culture platform liquid media, HyClone™ PF-CHO LS and PF-CHO MPS
Supplier: HyClone products (Cytiva)
HyClone™
SH30359.09EA 198.72 EUR
SH30359.09 HYCLSH30359.03 HYCLSH30359.02 SH30335.01 SH30334.01 HYCLSH30333.01 HYCLSH30333.02
CHO cell culture platform liquid media, HyClone™ PF-CHO LS and PF-CHO MPS
Cell Culture Media
HyClone™ PF-CHO LS and PF-CHO MPS media are designed to support the DHFR selection/amplification system. They have been successfully tested in a variety of cell culture systems, including T-flasks, spinner flasks, and bioreactors. PF-CHO MPS media is the powder version of PF-CHO LS media.
  • Designed to support the DHFR selection/amplification system
  • Successfully tested in a variety of cell culture systems, including T-flasks, spinner flasks and bioreactors
  • Enable growth of many CHO cells with minimal adaptation
  • PF-CHO LS liquid medium contains 4 mM L-glutamine and 0,1% poloxamer 188
  • PF-CHO MPS powder medium does not contain L-glutamine or poloxamer 188
HyClone™ PF-CHO LS and PF-CHO MPS media are ideally suited for the cost-effective manufacture of recombinant proteins for academic and industrial applications, genomics and proteomics, in vitro diagnostics, drug target screening and validation, and manufacturing of pre-clinical lots.