SFM4 insect media, HyClone™
Supplier: HyClone products (Cytiva)
HyClone™
SH30912.01EA 265 EUR
SH30912.01 SH30913.02 SH30913.03
SFM4 insect media, HyClone™
Cell Culture Media
HyClone™ SFM4 insect is a versatile cell culture medium developed through Metabolic Pathway Design to support the growth of multiple insect cell lines and production of a variety of recombinant proteins without compromising performance. HyClone SFM4 insect provides superior growth of many key insect cell lines, including Sf9, Sf21, High Five (T.ni), and D.mel.
  • Supports multiple insect cell lines
  • Free of animal derived components
  • Protein-free
  • High performance growth in multiple culture systems, including high density bioreactors
  • Full component traceability
  • Manufactured in cGMP and ISO compliant facility
Certifications: Manufactured using ISO 9001 and ISO 13485 certified processes according to 21 CFR 820 medical device current good manufacturing practices (cGMP).