Bovine fetuin (from Fetal serum)
Supplier: Merck
SIALF2379-1GEA 229 EUR
SIALF2379-1G SIALF2379-100MG SIALF2379-25G SIALF2379-250MG SIALF2379-5G
Bovine fetuin (from Fetal serum)
Proteins and Peptides
Fetuin extracted from foetal bovine serum. Fetuin is an α-globulin that inhibits trypsin activity and promotes cell attachment, growth and differentiation in many different culture systems.
  • Lyophilized powder
  • Impurities ≤0,3% free N-acetylneuraminic acid