Human N-Succinyl-Ala-Ala-Pro-Phe p-nitroanilide
Supplier: Merck
SIALS7388-100MGEA 419 EUR
SIALS7388-100MG SIALS7388-500MG SIALS7388-250MG SIALS7388-25MG
Human N-Succinyl-Ala-Ala-Pro-Phe p-nitroanilide
Proteins and Peptides
N-Succinyl-Ala-Ala-Pro-Phe p-nitroanilide is a colorimetric substrate for chymotrypsin, human leukocyte cathepsin G and peptidyl prolyl isomerase.