Healthy human chondrocytes P0 (50-70y), CELLvo™
Supplier: StemBioSys
STBSHCA3-100F-T75EA 1410 EUR
STBSHCA3-100F-T75 STBSHCA3-100M-T150 STBSHCA3-100M-T75 STBSHCA3-100F-T150 STBSHCA3-100M-000 STBSHCA3-100M-6WP STBSHCA3-100F-6WP STBSHCA3-100F-000
Healthy human chondrocytes P0 (50-70y), CELLvo™
Cells
CELLvo™ Healthy Human Chondrocytes (50-70y) are primary, uncultured cells isolated from recently-diseased cadaver cartilage from donors between 50 and 70 years old. Healthy status is determined by macroscopic inspection of the tissue using a modified Outerbridge (1960) scale in order to distinguish healthy joints from those with signs of osteoarthritis. Cells are isolated by enzymatic digestion prior to cryopreservation.