Enclosed PPE dispensers
Supplier: Teknomek
TEKMDPPE-GEA 0 EUR
TEKMDPPE-G TEKMDPPE-GF TEKMHWDI-MG4S TEKMDPPE-G4F
Enclosed PPE dispensers
PPE Dispensers
Our enclosed PPE dispenser offers intelligent design for all your PPE needs with an easy-clean sloping top.
  • Easy to clean intelligent design
  • Sloping top
  • Design has no plastic parts
  • Available in 316-grade stainless steel
  • Brushed dull polish finish
  • With or without flaps
  • Wall fixed - option to add magnets for no damage wall fixing
Our glove box and face mask dispenser is intelligently designed as it comes with no plastic parts and with a sloping top that makes it very easy to clean.
The glove box/face mask dispenser is manufactured in 304-grade stainless steel for ultimate hygiene and durability, with a 316-grade option also available for optimised toughness and corrosion resistance.
The enclosed PPE dispenser comes in a brushed dull polish finish. It comes with a sloping top for ideal storage and are wall-fixed to ensure full space efficiency.
The range is easy for boxes of a wide range of sizes to quickly be installed, or for the 4 tier designs, you can opt to be 100% stainless steel by selecting our glove holder as an insert - DPPE-ACGI.
The dispensers are suited to rigorous hygiene and HACCP routines, meaning you can rely on us to deliver a hygienic, durable and easy to clean product ideal for your workspace.