Adjustable solid wall shelving
Supplier: Teknomek
TEKMMWA0400/1200/3EA 0 EUR
TEKMMWA0400/1200/3 TEKMMWA0300/1500/2 TEKMMWA0300/0900/3 TEKMMWA0300/0900/2 TEKMMWA0400/1200/2 TEKMMWA0400/0900/2 TEKMMWA0300/1800/2 TEKMMWA0300/1800/3 TEKMMWA0300/1000/3 TEKMMWA0300/1000/2 TEKMMWA0300/1200/3 TEKMMWA0400/0900/3 TEKMMWA0300/1200/2 TEKMMWA0400/1800/2 TEKMMWA0400/1800/3 TEKMMWA0400/1500/3 TEKMMWA0400/1000/2 TEKMMWA0400/1500/2 TEKMMWA0400/1000/3 TEKMMWA0300/1500/3
Adjustable solid wall shelving
Furniture Shelves
This adjustable wall shelving unit is made from 304 grade stainless steel and supplied with stainless brackets ensuring that this product is steady and tough.
  • Brushed dull polish finish
  • Fully adjustable shelves
  • Wall fixed
With flexible mounting positions and adjustments every 30 mm, you are sure to find shelves suitable for your needs.