Wash troughs
Supplier: Teknomek
TEKMWTN020EXBATPEDEA 0 EUR
TEKMWTN020EXBATPED TEKMWTN020EXBAT TEKMWTN040EXSEN TEKMWTN020EXSEN TEKMWTN010EX TEKMWTN050EXPED TEKMWTN030EX TEKMWTN060EXSENPED TEKMWTN060EX TEKMWTN040EXPED TEKMWTN050EX TEKMWTN050EXBAT TEKMWTN050EXSEN TEKMWTN010EXSENPED TEKMWTN020EXPED TEKMWTN040EXSENPED TEKMWTN010EXPED TEKMWTN020EXSENPED TEKMWTN030EXBAT TEKMWTN040EXBATPED TEKMWTN010EXBATPED TEKMWTN020EX TEKMWTN030EXSEN TEKMWTN040EX TEKMWTN010EXSEN TEKMWTN030EXSENPED TEKMWTN060EXSEN TEKMWTN060EXPED TEKMWTN010EXBAT TEKMWTN030EXPED TEKMWTN060EXBAT
Wash troughs
Faucets and Sinks Sinks
Our range of sensor operated wash troughs are manufactured from 304 grade stainless steel with a fully welded design for maximum hygiene. Choose from knee, battery or mains operation to suit your needs.
  • Secret wall fixing to hang the wash trough to to the wall
  • Supplied fitted, ready to plumb in
  • Infra-red sensor operated
  • Sloped trough for fast draining
Our wash troughs are manufactured in fully welded 304-grade stainless steel and can be wall-mounted or with additional pedestal support.
Being hands-free, the design supports rigorous hygienic standards.
Ordering information: Optional skirt (to be ordered alongside the wash trough as cannot be retrofitted).