Clipboards
Supplier: Teknomek
TEKMWW0200SSCCLIPEA 0 EUR
TEKMWW0200SSCCLIP TEKMWW0205LSSCCLIP TEKMWW0205/SSC TEKMWW0200SSCCLIPL TEKMWW0205/SSC/L TEKMWW0200/SSC/L TEKMWW0205SSCCLIP TEKMWW0200/SSC
Clipboards
Clipboards
The clipboards come in a range of options.
  • Brushed dull polish finish
  • Spring loaded stainless steel clip
  • Available in portrait or landscape
  • Material: Fully welded 304-grade stainless steel
The stainless steel clipboards come in sizes A4 and A3, and can be supplied with or without a clip at the back to hang them from our hanging rails.