Bottles, aluminium, with thread, SYSTEM PLUS® 45
Supplier: Tournaire
TOUR5148S1EA 1820 EUR
TOUR5148S1 TOUR5376S1 TOUR5220S1 TOUR7418S1 TOUR4829S1 TOUR5453S1 TOUR7419S1 TOUR4348S1 TOUR5454S1 TOUR7858S1 TOUR5459S1 215-3153
Bottles, aluminium, with thread, SYSTEM PLUS® 45
Bottles Metal Bottles
Made of pure aluminium.
  • Unbreakable, odorless
  • Very good temperature resistance
  • Leakproof with tamper-evident screw caps (must be odered separately)
  • UN-approved
Suitable for perfumes compositions, aroma chemicals, food ingredients and laboratory liquid reagents.